Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdgC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001370029.1 | NP_001356983.1 | 6 - 7 | 1032 | 63 | |
NM_080490.4 | NP_536738.3 | 7 - 8 | 1881 | 63 | ||
NM_176366.2 | NP_788544.2 | 7 - 8 | 1519 | 63 | ||
NM_176367.3 | NP_788545.3 | 7 - 8 | 1694 | 63 | ||
NM_176368.2 | NP_788546.1 | 6 - 7 | 1209 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |