Search
Search

Gprk2| Gene Symbol: Gprk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276214.1 | NP_001263143.1 | 7 - 8 | 2261 | 89 | |
| NM_057519.4 | NP_476867.1 | 8 - 9 | 2309 | 89 | ||
| GenBank mRNA | BT031258.1 | - | 8 - 9 | 1773 | 89 | |
Set Membership: |
|