Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm02134721_m1
          See other TWS GE Assays ›
          Gene Symbol:
          tws
          Gene Name:
          twins
          Gene Aliases:
          1466/06, 2414, B/PR55, CG6235, DPR55, Dmel\CG6235, Dmel_CG6235, PP2A, PP2A-B55, PP2A[B], PP2a, PR55, Pp2A-85F, Twins, Tws, aar, dPP2A, dPP2A-B55, dPP2A-Balpha, dPR55, l(3)01436, l(3)02414, l(3)S022205b, l(3)S022361, l(3)S022440, l(3)S023013, l(3)S023141a, l(3)S023206, l(3)S023309, l(3)S023938, l(3)S024455, l(3)S024834a, l(3)S024838, l(3)S025806, l(3)S025832, l(3)S025913, l(3)S026226, l(3)S026326, l(3)S027127, l(3)S027313, l(3)S028707, l(3)S029110, l(3)S029403, l(3)S029701a, l(3)S031006, l(3)S032303, l(3)S032708c, l(3)S033903, l(3)S035505b, l(3)S042629, l(3)S042630, l(3)S043008b, l(3)S043029, l(3)S045519, l(3)S046918, l(3)S048006, l(3)S048013, l(3)S048507, l(3)S049902, l(3)S052810, l(3)S053011, l(3)S060203, l(3)S060804, l(3)S061805, l(3)S061915, l(3)S063110, l(3)S066017, l(3)S066813, l(3)S067109b, l(3)S067915, l(3)S069206a, l(3)S075110, l(3)S075515b, l(3)S075902b, l(3)S076415, l(3)S080409, l(3)S088513, l(3)S091905, l(3)S101413b, l(3)S105605, l(3)S110008a, l(3)S110815, l(3)S111515, l(3)S119908, l(3)S132907, l(3)S134601a, l(3)S141309, l(3)S146606, l(3)j11C8, l(3)s1801, tw, tws/aar, v158
          Chromosome Location:
          Chr.3R: 10126129 - 10139772 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0004889
          Assay ID Dm02134721_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: tws
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001300340.1 NP_001287269.1 1 - 2 512 81
          NM_057532.4 NP_476880.1 2 - 3 691 81
          NM_169329.2 NP_731451.1 2 - 3 320 81
          GenBank mRNA AY061152.1 - 2 - 3 667 81
          L07581.1 - 2 - 3 525 81

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          protein dephosphorylation
          metaphase/anaphase transition of mitotic cell cycle
          centrosome cycle
          negative regulation of neuroblast proliferation
          sensory organ development
          imaginal disc pattern formation
          regulation of mitotic metaphase/anaphase transition
          maintenance of neuroblast polarity
          positive regulation of smoothened signaling pathway
          regulation of centriole replication
          protein stabilization
          peptidyl-serine dephosphorylation
          positive regulation of canonical Wnt signaling pathway
          protein phosphatase regulator activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline