Search
Search

cno| Gene Symbol: cno | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275342.1 | NP_001262271.1 | 8 - 9 | 2333 | 69 | |
| NM_001275343.1 | NP_001262272.1 | 11 - 12 | 3313 | 69 | ||
| NM_001275344.1 | NP_001262273.1 | 13 - 14 | 3613 | 69 | ||
| NM_001275345.1 | NP_001262274.1 | 13 - 14 | 3613 | 69 | ||
| NM_001316565.1 | NP_001303494.1 | 13 - 14 | 3613 | 69 | ||
| NM_079508.4 | NP_524232.2 | 13 - 14 | 3613 | 69 | ||
| NM_169025.3 | NP_730891.1 | 10 - 11 | 3101 | 69 | ||
| NM_169026.3 | NP_730892.2 | 13 - 14 | 3613 | 69 | ||
| GenBank mRNA | BT044227.1 | - | 13 - 14 | 3582 | 69 | |
Set Membership: |
|