Search
Search

Rm62| Gene Symbol: Rm62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170057.1 | NP_001163528.1 | 3 - 4 | 1409 | 71 | |
| NM_001202253.2 | NP_001189182.1 | 3 - 4 | 1797 | 71 | ||
| NM_001260009.1 | NP_001246938.1 | 3 - 4 | 1401 | 71 | ||
| NM_001260010.1 | NP_001246939.1 | 3 - 4 | 1406 | 71 | ||
| NM_001260011.1 | NP_001246940.1 | 3 - 4 | 1406 | 71 | ||
| NM_001260012.1 | NP_001246941.1 | 3 - 4 | 1401 | 71 | ||
| NM_079519.2 | NP_524243.2 | 2 - 3 | 1644 | 71 | ||
| NM_169118.1 | NP_731031.1 | 3 - 4 | 1406 | 71 | ||
| NM_169119.3 | NP_731032.1 | 3 - 4 | 1809 | 71 | ||
| NM_169120.1 | NP_731033.1 | 4 - 5 | 1498 | 71 | ||
| NM_169121.2 | NP_731034.1 | 3 - 4 | 1482 | 71 | ||
| NM_169122.1 | NP_731035.2 | 3 - 4 | 1736 | 71 | ||
| GenBank mRNA | BT001716.1 | - | 4 - 5 | 1492 | 71 | |
| BT011476.1 | - | 2 - 3 | 1644 | 71 | ||
| BT015209.1 | - | 3 - 4 | 1406 | 71 | ||
| BT099694.1 | - | 3 - 4 | 1527 | 71 | ||
Set Membership: |
|