Search
Search

svp| Gene Symbol: svp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275570.1 | NP_001262499.1 | 3 - 4 | 1799 | 82 | |
| NM_001382014.1 | NP_001369011.1 | 3 - 4 | 1799 | 82 | ||
| NM_079601.3 | NP_524325.1 | 3 - 4 | 1799 | 82 | ||
| NM_169459.2 | NP_731682.1 | 3 - 4 | 816 | 82 | ||
| GenBank mRNA | AY075272.1 | - | 3 - 4 | 804 | 82 | |
| M28863.1 | - | 3 - 4 | 1806 | 82 | ||
Set Membership: |
|