Search
Search

sr| Gene Symbol: sr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170172.2 | NP_001163643.1 | 2 - 3 | 1183 | 89 | |
| NM_001275764.1 | NP_001262693.1 | 4 - 5 | 2397 | 89 | ||
| NM_001300455.1 | NP_001287384.1 | 2 - 3 | 1183 | 89 | ||
| NM_079671.2 | NP_524395.2 | 4 - 5 | 2397 | 89 | ||
| NM_169786.3 | NP_732289.1 | 2 - 3 | 1183 | 89 | ||
| GenBank mRNA | BT133222.1 | - | 2 - 3 | 1178 | 89 | |
Set Membership: |
|