Search
Search

fru| Gene Symbol: fru | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170177.2 | NP_001163648.1 | 4 - 5 | 936 | 55 | |
| NM_001275783.1 | NP_001262712.1 | 3 - 4 | 2522 | 55 | ||
| NM_079673.3 | NP_524397.2 | 3 - 4 | 2522 | 55 | ||
| NM_169816.2 | NP_732344.1 | 3 - 4 | 921 | 55 | ||
| NM_169817.2 | NP_732345.1 | 3 - 4 | 921 | 55 | ||
| NM_169818.2 | NP_732346.1 | 3 - 4 | 2522 | 55 | ||
| NM_169819.2 | NP_732347.1 | 3 - 4 | 921 | 55 | ||
| NM_169820.2 | NP_732348.1 | 3 - 4 | 2522 | 55 | ||
| NM_169821.2 | NP_732349.1 | 2 - 3 | 784 | 55 | ||
| NM_169822.2 | NP_732350.1 | 2 - 3 | 175 | 55 | ||
| NM_206512.2 | NP_996234.1 | 2 - 3 | 227 | 55 | ||
| NM_206513.2 | NP_996235.1 | 2 - 3 | 784 | 55 | ||
| NM_206514.2 | NP_996236.1 | 3 - 4 | 773 | 55 | ||
| NM_206515.2 | NP_996237.1 | 2 - 3 | 376 | 55 | ||
| NM_206516.2 | NP_996238.1 | 2 - 3 | 224 | 55 | ||
| GenBank mRNA | BT024450.1 | - | 4 - 5 | 936 | 55 | |
Set Membership: |
|