Search
Search

5-HT2B| Gene Symbol: 5-HT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275444.2 | NP_001262373.1 | 5 - 6 | 1651 | 85 | |
| NM_001300309.1 | NP_001287238.1 | 5 - 6 | 1651 | 85 | ||
| NM_141549.4 | NP_649806.2 | 5 - 6 | 1651 | 85 | ||
| GenBank mRNA | KC852205.1 | - | 4 - 5 | 1008 | 85 | |
Set Membership: |
|