Search
Search

rdx| Gene Symbol: rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142068.2 | NP_650325.1 | 4 - 5 | 646 | 137 | |
| NM_142069.4 | NP_650326.3 | 3 - 4 | 2003 | 137 | ||
| NM_169564.2 | NP_731875.2 | 3 - 4 | 806 | 137 | ||
| NM_169565.2 | NP_731876.2 | 3 - 4 | 806 | 137 | ||
| NM_169566.2 | NP_731877.1 | 4 - 5 | 825 | 137 | ||
| GenBank mRNA | AY121682.1 | - | 4 - 5 | 823 | 137 | |
| BT012443.1 | - | 4 - 5 | 636 | 137 | ||
| BT021321.1 | - | 3 - 4 | 2002 | 137 | ||
| BT099706.1 | - | 3 - 4 | 804 | 137 | ||
Set Membership: |
|