Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acyp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275683.1 | NP_001262612.1 | 3 - 4 | 341 | 70 | |
NM_142234.2 | NP_650491.1 | 3 - 4 | 341 | 70 | ||
GenBank mRNA | BT024404.1 | - | 3 - 4 | 336 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |