Search
Search

CG5903| Gene Symbol: CG5903 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300427.1 | NP_001287356.1 | 1 - 2 | 320 | 89 | |
| NM_001316507.1 | NP_001303436.1 | 1 - 2 | 433 | 89 | ||
| NM_142266.3 | NP_650523.1 | 1 - 2 | 433 | 89 | ||
| GenBank mRNA | AY060852.1 | - | 1 - 2 | 433 | 89 | |
| BT021237.1 | - | 1 - 2 | 267 | 89 | ||
Set Membership: |
|