Search
Search

Det| Gene Symbol: Det | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_142351.4 | NP_650608.1 | 2 - 3 | 435 | 63 | |
| GenBank mRNA | AY071487.1 | - | 2 - 3 | 408 | 63 | |
| KX532033.1 | - | 2 - 3 | 331 | 63 | ||
Set Membership: |
|