Search
Search

Brf| Gene Symbol: Brf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170165.2 | NP_001163636.1 | 1 - 2 | 245 | 62 | |
| NM_142359.3 | NP_650616.2 | 1 - 2 | 245 | 62 | ||
| GenBank mRNA | AY051811.1 | - | 1 - 2 | 236 | 62 | |
Set Membership: |
|