Search
Search

dsx| Gene Symbol: dsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275423.1 | NP_001262352.1 | 3 - 4 | 2065 | 57 | |
| NM_001275424.1 | NP_001262353.1 | 3 - 4 | 2454 | 57 | ||
| NM_169202.1 | NP_731197.1 | 3 - 4 | 2454 | 57 | ||
Set Membership: |
|