Search
Search

AdamTS-A| Gene Symbol: AdamTS-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275675.1 | NP_001262604.1 | 11 - 12 | 3440 | 63 | |
| NM_169659.3 | NP_732036.2 | 11 - 12 | 3208 | 63 | ||
| NM_206496.2 | NP_996218.1 | 11 - 12 | 4241 | 63 | ||
| GenBank mRNA | AY094716.1 | - | 11 - 12 | 4240 | 63 | |
Set Membership: |
|