Search
Search

tinc| Gene Symbol: tinc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275757.1 | NP_001262686.1 | 5 - 6 | 1404 | 75 | |
| NM_169780.5 | NP_732278.2 | 6 - 7 | 1491 | 75 | ||
| NM_176509.4 | NP_788686.1 | 6 - 7 | 1491 | 75 | ||
| NM_176510.5 | NP_788687.3 | 6 - 7 | 1401 | 75 | ||
| NM_176511.3 | NP_788688.2 | 6 - 7 | 1449 | 75 | ||
| GenBank mRNA | AF509933.1 | - | 3 - 4 | 666 | 75 | |
Set Membership: |
|