Search
Search

| Gene Symbol: CG33293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206428.2 | NP_996150.1 | 2 | 536 | 128 | |
| GenBank mRNA | AY060254.1 | - | 1 | 3185 | 128 | |
| BT014674.1 | - | 2 | 536 | 128 | ||
| Gene Symbol: CG12163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY121614.1 | - | 2 | 4384 | 128 | |
Set Membership: |
|