Search
Search

Dpy-30L2| Gene Symbol: Dpy-30L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274464.1 | NP_001261393.1 | 2 | 427 | 81 | |
| NM_139597.2 | NP_647854.1 | 2 | 471 | 81 | ||
| GenBank mRNA | BT023602.1 | - | 2 | 449 | 81 | |
Set Membership: |
|