Search
Search

nitr2b| Gene Symbol: nitr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131649.2 | NP_571724.1 | 5 - 6 | 960 | 146 | |
| XM_017356909.1 | XP_017212398.1 | 5 - 6 | 955 | 146 | ||
| GenBank mRNA | AF318395.1 | - | 4 - 5 | 818 | 146 | |
| AF318396.1 | - | 4 - 5 | 818 | 146 | ||
| AF318397.1 | - | 4 - 5 | 818 | 146 | ||
| BC162753.1 | - | 4 - 5 | 869 | 146 | ||
| BC162759.1 | - | 4 - 5 | 869 | 146 | ||
| CU638755.1 | - | 4 - 5 | 818 | 146 | ||
Set Membership: |
|