Search
Search

fgf20a| Gene Symbol: fgf20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037103.2 | NP_001032180.1 | 2 - 3 | 474 | 64 | |
| GenBank mRNA | BC162861.1 | - | 2 - 3 | 474 | 64 | |
| BC162886.1 | - | 2 - 3 | 474 | 64 | ||
| DQ195158.1 | - | 2 - 3 | 381 | 64 | ||
Set Membership: |
|