Search
Search

crygm2a| Gene Symbol: crygm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018121.1 | NP_001018131.1 | 1 - 2 | 70 | 60 | |
| GenBank mRNA | BC163158.1 | - | 1 - 2 | 66 | 60 | |
| BC163189.1 | - | 1 - 2 | 66 | 60 | ||
| DQ009661.1 | - | 1 - 2 | 70 | 60 | ||
Set Membership: |
|