Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131152.1 | NP_571227.1 | 2 - 3 | 130 | 58 | |
GenBank mRNA | AF032392.1 | - | 2 - 3 | 130 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |