Search
Search

caspb| Gene Symbol: caspb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152884.2 | NP_690840.2 | 2 - 3 | 312 | 71 | |
| GenBank mRNA | AF327410.1 | - | 1 - 2 | 275 | 71 | |
| BC095000.1 | - | 2 - 3 | 312 | 71 | ||
| BC164498.1 | - | 1 - 2 | 309 | 71 | ||
| DQ022755.1 | - | 1 - 2 | 275 | 71 | ||
| MG958005.1 | - | 1 - 2 | 275 | 71 | ||
Set Membership: |
|