Search
Search

rxrba| Gene Symbol: rxrba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131275.1 | NP_571350.1 | 12 - 13 | 1447 | 94 | |
| XM_005159410.3 | XP_005159467.1 | 11 - 12 | 1446 | 94 | ||
| XM_005159411.3 | XP_005159468.1 | 10 - 11 | 1422 | 94 | ||
| XM_005159412.3 | XP_005159469.1 | 9 - 10 | 1380 | 94 | ||
| XM_009294022.2 | XP_009292297.1 | 12 - 13 | 1473 | 94 | ||
| XM_009294023.2 | XP_009292298.1 | 11 - 12 | 1449 | 94 | ||
| XM_009294024.2 | XP_009292299.1 | 11 - 12 | 1431 | 94 | ||
| XM_009294025.2 | XP_009292300.1 | 10 - 11 | 1407 | 94 | ||
| XM_009294026.2 | XP_009292301.1 | 12 - 13 | 1495 | 94 | ||
| XM_021467983.1 | XP_021323658.1 | 13 - 14 | 1522 | 94 | ||
| GenBank mRNA | BC054649.1 | - | 11 - 12 | 1272 | 94 | |
| U29942.1 | - | 12 - 13 | 1447 | 94 | ||
Set Membership: |
|