Search
Search

rps9| Gene Symbol: rps9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200852.3 | NP_957146.1 | 4 - 5 | 459 | 107 | |
| GenBank mRNA | BC059492.1 | - | 3 - 4 | 441 | 107 | |
| BC062833.1 | - | 3 - 4 | 443 | 107 | ||
| CT730686.2 | - | 3 - 4 | 442 | 107 | ||
Set Membership: |
|