Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03150153_m1
          See other CDH2 GE Assays ›
          Gene Symbol:
          cdh2
          Gene Name:
          cadherin 2, type 1, N-cadherin (neuronal)
          Gene Aliases:
          bib, glo, lyr, ncad, pac, wu:fb47h04
          Chromosome Location:
          Chr.20: 17740104 - 17819238 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03150153_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: cdh2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_131081.2 NP_571156.2 5 - 6 814 63
          GenBank mRNA AF418565.1 - 5 - 6 810 63
          AF430842.1 - 5 - 6 809 63
          BC133731.1 - 5 - 6 797 63
          KM655603.1 - 5 - 6 665 63
          X67648.1 - 351 63

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          cartilage condensation
          neural crest cell migration
          somitogenesis
          neuron migration
          cell adhesion
          homophilic cell adhesion via plasma membrane adhesion molecules
          neuron cell-cell adhesion
          multicellular organism development
          ectoderm development
          nervous system development
          axon guidance
          axonal fasciculation
          synapse assembly
          brain development
          peripheral nervous system development
          motor neuron axon guidance
          regulation of signal transduction
          glial cell differentiation
          cardioblast differentiation
          spinal cord patterning
          cell migration in hindbrain
          hindbrain structural organization
          ventricular system development
          neural tube development
          cell differentiation
          midbrain development
          hindbrain development
          adherens junction organization
          embryonic pectoral fin morphogenesis
          dorsal fin morphogenesis
          post-anal tail morphogenesis
          tissue regeneration
          odontogenesis
          regulation of eye photoreceptor cell development
          cell-cell adhesion mediated by cadherin
          establishment of epithelial cell apical/basal polarity
          embryonic eye morphogenesis
          axial mesoderm structural organization
          mesoderm morphogenesis
          blood vessel morphogenesis
          generation of neurons
          semicircular canal morphogenesis
          regulation of axonogenesis
          regulation of synaptic transmission, glutamatergic
          dorsal convergence
          retina development in camera-type eye
          embryonic retina morphogenesis in camera-type eye
          auditory receptor cell stereocilium organization
          cilium morphogenesis
          pigment cell development
          sensory epithelium regeneration
          commissural neuron axon guidance
          cell migration involved in somitogenic axis elongation
          neuronal stem cell population maintenance
          motor neuron migration
          cell-cell adhesion
          cell-cell adhesion via plasma-membrane adhesion molecules
          negative regulation of neural precursor cell proliferation
          calcium ion binding
          cadherin binding
          metal ion binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0