Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006100.1 | NP_001006100.1 | 3 - 4 | 415 | 142 | |
XM_009307310.3 | XP_009305585.1 | 4 - 5 | 352 | 142 | ||
XM_017358735.2 | XP_017214224.1 | 4 - 5 | 514 | 142 | ||
GenBank mRNA | BC083216.1 | - | 3 - 4 | 415 | 142 | |
BC164088.1 | - | 3 - 4 | 348 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |