Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: valopb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110280.1 | NP_001103750.1 | 3 - 4 | 756 | 90 | |
GenBank mRNA | AY996588.1 | - | 3 - 4 | 756 | 90 | |
KT008403.1 | - | 3 - 4 | 674 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Danio rerio Probe spans exons |