Search
Search

saga| Gene Symbol: saga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200559.2 | NP_956853.1 | 9 - 10 | 810 | 90 | |
| GenBank mRNA | BC056570.1 | - | 757 | 90 | ||
| BC068418.1 | - | 751 | 90 | |||
Set Membership: |
|