Search
Search

hpcal4| Gene Symbol: hpcal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114687.1 | NP_001108159.1 | 1 - 2 | 161 | 75 | |
| XM_005159639.4 | XP_005159696.1 | 2 - 3 | 318 | 75 | ||
Set Membership: |
|