Search
Search

cxcl32b.1| Gene Symbol: cxcl32b.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115061.1 | NP_001108533.1 | 1 - 2 | 77 | 82 | |
| GenBank mRNA | AB331759.1 | - | 1 - 2 | 77 | 82 | |
Set Membership: |
|