Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201118.2 | NP_957412.2 | 8 - 9 | 991 | 72 | |
GenBank mRNA | BC055598.1 | - | 8 - 9 | 864 | 72 | |
BC150464.1 | - | 8 - 9 | 850 | 72 | ||
BC165143.1 | - | 5 - 6 | 480 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |