Search
Search

gpm6aa| Gene Symbol: gpm6aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213200.1 | NP_998365.1 | 2 - 3 | 335 | 102 | |
| GenBank mRNA | AY070261.1 | - | 2 - 3 | 335 | 102 | |
| BC064281.1 | - | 2 - 3 | 276 | 102 | ||
| BC150455.1 | - | 2 - 3 | 359 | 102 | ||
Set Membership: |
|