Search
Search
| Gene Symbol: GJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081891.1 | NP_001075360.1 | 1 | 49 | 60 | |
| XM_005614219.3 | XP_005614276.1 | 177 | 60 | |||
| XM_005614221.3 | XP_005614278.1 | 149 | 60 | |||
| XM_023633177.1 | XP_023488945.1 | 168 | 60 | |||
| GenBank mRNA | AY286490.1 | - | 1 | 49 | 60 | |
Set Membership: |
|