Search
Search

PMP22| Gene Symbol: PMP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081936.1 | NP_001075405.1 | 1 - 2 | 113 | 70 | |
| XM_005597020.2 | XP_005597077.1 | 400 | 70 | |||
| XM_005597021.3 | XP_005597078.1 | 286 | 70 | |||
| GenBank mRNA | AY280708.1 | - | 1 - 2 | 113 | 70 | |
Set Membership: |
|