Search
Search

RBP4
LOC111773844| Gene Symbol: RBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081951.1 | NP_001075420.1 | 3 - 4 | 527 | 61 | |
| XM_023655349.1 | XP_023511117.1 | 395 | 61 | |||
| GenBank mRNA | U21208.1 | - | 3 - 4 | 527 | 61 | |
| Gene Symbol: LOC111773844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023642359.1 | XP_023498127.1 | 603 | 61 | ||
| XM_023642368.1 | XP_023498136.1 | 395 | 61 | |||
Set Membership: |
|