Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256899.1 | NP_001243828.1 | 1 | 594 | 82 | |
XM_014738512.2 | XP_014593998.2 | 756 | 82 | |||
GenBank mRNA | EF581164.1 | - | 1 | 252 | 82 | |
JL624769.1 | - | 2 | 758 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Equus caballus Both primers and probe map within a single exon |