Search
Search

PHF5A| Gene Symbol: PHF5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309325.1 | NP_001296254.1 | 2 - 3 | 86 | 62 | |
| GenBank mRNA | CX597298.1 | - | 2 - 3 | 87 | 62 | |
| JL616446.1 | - | 2 - 3 | 106 | 62 | ||
Set Membership: |
|