Search
Search

FERMT2| Gene Symbol: FERMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936527.3 | XP_004936584.2 | 12 - 13 | 1534 | 76 | |
| XM_025150867.1 | XP_025006635.1 | 11 - 12 | 1422 | 76 | ||
| XM_025150868.1 | XP_025006636.1 | 10 - 11 | 1389 | 76 | ||
| XM_025150869.1 | XP_025006637.1 | 10 - 11 | 1528 | 76 | ||
| GenBank mRNA | CR385726.1 | - | 7 - 8 | 695 | 76 | |
Set Membership: |
|