Search
Search

COL3A1| Gene Symbol: COL3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205380.2 | NP_990711.2 | 33 - 34 | 2474 | 109 | |
| XM_015289138.2 | XP_015144624.1 | 37 - 38 | 2863 | 109 | ||
| XM_025152154.1 | XP_025007922.1 | 32 - 33 | 2430 | 109 | ||
| GenBank mRNA | AM064821.1 | - | 9 - 10 | 541 | 109 | |
| L31344.1 | - | 12 - 13 | 797 | 109 | ||
| U07973.1 | - | 33 - 34 | 2451 | 109 | ||
| U07974.1 | - | 12 - 13 | 797 | 109 | ||
Set Membership: |
|