Search
Search

SLC22A16| Gene Symbol: SLC22A16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940304.3 | XP_004940361.1 | 2 - 3 | 1920 | 65 | |
| XM_015284571.2 | XP_015140057.1 | 3 - 4 | 776 | 65 | ||
| XM_025149130.1 | XP_025004898.1 | 2 - 3 | 3949 | 65 | ||
| XM_419787.6 | XP_419787.3 | 2 - 3 | 571 | 65 | ||
| GenBank mRNA | BX931814.1 | - | 2 - 3 | 526 | 65 | |
| CR353847.1 | - | 3 - 4 | 622 | 65 | ||
Set Membership: |
|