Search
Search

TMEM26| Gene Symbol: TMEM26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199598.1 | NP_001186527.1 | 4 - 5 | 742 | 74 | |
| GenBank mRNA | BX932258.2 | - | 4 - 5 | 457 | 74 | |
| BX933778.2 | - | 1 - 2 | 91 | 74 | ||
| BX950714.1 | - | 4 - 5 | 742 | 74 | ||
Set Membership: |
|