Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MYLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012561.1 | NP_001012579.1 | 6 - 7 | 1386 | 86 | |
GenBank mRNA | AJ851471.1 | - | 6 - 7 | 1386 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |