Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205354.1 | NP_990685.1 | 1 - 2 | 215 | 143 | |
XM_015286041.2 | XP_015141527.1 | 1 - 2 | 860 | 143 | ||
GenBank mRNA | U20615.1 | - | 1 - 2 | 215 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons |