Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IRF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031587.1 | NP_001026758.1 | 5 - 6 | 615 | 97 | |
XM_025151342.1 | XP_025007110.1 | 5 - 6 | 978 | 97 | ||
XM_025151344.1 | XP_025007112.1 | 4 - 5 | 2045 | 97 | ||
GenBank mRNA | AJ720409.1 | - | 5 - 6 | 615 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |