Search Thermo Fisher Scientific
Gene Symbol: PECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006522.1 | NP_001006522.1 | 2 - 3 | 264 | 83 | |
GenBank mRNA | AJ720820.1 | - | 2 - 3 | 264 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |