Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006577.2 | NP_001006577.2 | 10 - 11 | 1038 | 88 | |
XM_015277311.2 | XP_015132797.1 | 11 - 12 | 1073 | 88 | ||
GenBank mRNA | AJ720892.1 | - | 10 - 11 | 1043 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |