Search
Search

NECAP1| Gene Symbol: NECAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640439.4 | XP_003640487.1 | 7 - 8 | 879 | 85 | |
| GenBank mRNA | BX932228.1 | - | 4 - 5 | 439 | 85 | |
| BX932268.1 | - | 3 - 4 | 368 | 85 | ||
| BX935136.1 | - | 5 - 6 | 437 | 85 | ||
| CR389569.1 | - | 7 - 8 | 585 | 85 | ||
Set Membership: |
|