Search
Search

SUT1| Gene Symbol: SUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249369.1 | NP_001236298.1 | 1 - 2 | 1292 | 66 | |
| GenBank mRNA | AJ563364.1 | - | 1 - 2 | 1292 | 66 | |
| AK244954.1 | - | 1 - 2 | 1338 | 66 | ||
Set Membership: |
|